Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ SNaPshot™ Multiplex Kit

Catalog No. p-7138597
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
100 Reactions
100 Reactions, with protocol
1000 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
43-231-59 100 Reactions
43-231-51 100 Reactions, with protocol
43-231-61 1000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 43-231-59 Supplier Applied Biosystems™ Supplier No. 4323159
Only null left
Add to Cart
Edge
Add to Cart

Includes

SNaPshot™ Multiplex Ready Reaction Mix (for 100 reactions), SNaPshot™ Multiplex Control Primer Mix (30μL), SNaPshot™ Multiplex Control Template (60μL)

The SNaPshot™ Multiplex Kit is part of the SNaPshot™ Multiplex System. The SNaPshot™ Multiplex System primer extension-based method enables multiplexing up to 10 single nucleotide polymorphisms (SNPs).

  • Interrogate up to 10 SNPs from different amplicons in a single base extension reaction
  • Separate SNP loci that differ by a single base pair
  • Increased sensitivity compared to standard sequencing allows detection of minor variants (e.g. somatic mutations) in a test sample
  • Reduce the complexity of your experiments with this easy to use and cost-efficient genotyping system
  • A large body of scientific literature demonstrates the utility for genotyping, mutation detection and mapping

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains:
• SNaPshot™ Multiplex Ready Reaction Mix (for 100 reactions)
• SNaPshot™ Multiplex Control Primer Mix (30 μl)
• SNaPshot™ Multiplex Control Template (60 μl)

Store at -15°C to -25°C in a constant-temperature freezer.
Description SNaPshot™ Multiplex Kit, 100 rxns, Capillary Electrophoresis Detection Method, DNA Sample Type, AmpliTaq™ DNA Polymerase, SNPs (Known) Genotyping Target, Multiplexing High Throughput Compatibility, Unlabelled Oligos Primer-Probe Compatibility
Detection Method Primer-probe
Format Tube
Polymerase AmpliTaq DNA Polymerase
Reaction Speed 8 hr.
Technique SNP Genotyping (Fragment Analysis-Based), Primer Extension
For Use With (Equipment) 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer, 3730/3730xl Genetic Analyzers
High-throughput Compatibility Multiplexing
Product Line SNaPshot
Product Type Multiplex Kit
Quantity 100 reactions
Run Time 8 hr.
Shipping Condition Dry Ice
Software GeneMapper™ Software
Species All Species
Target SNPs (Known)
For Use With (Application) Genotyping
Hot Start No
No. of Reactions 100 Reactions
Sample Type DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.