Learn More
Innovative Research Inc Sngl Dnr Hmn Diabetes Pla
Supplier:  Innovative Research Inc IPLASDIB
Encompass_Preferred
Single Donor Human Diabetes Plasma - Single Donor Human Diabetes Plasma from Innovative Research is whole blood derived. Available volumes are variable, and range from 0.5ml to 1.0ml per vial (you can select your exact samples from our inventory, if desired). A detailed patient history and clinical information is recorded at the time of draw, and multiple samples and plasma/serum matched sets from the same donor are often available.
Catalog No. 50-274-4107
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.