Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNRPA Antibody, ProSci
SDP

Catalog No. 16816648
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-816-648 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-816-648 Supplier ProSci LLC Supplier No. 222000100
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SNRPA Polyclonal Antibody specifically detects CCBL1 in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.

Specifications

Antigen SNRPA
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution WB: 1:500 - 1:2000, IHC: 1:50 - 1:200, IF: 1:50 - 1:200
Formulation PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
Gene Alias SNRPA, U1A, Mud1, U1-A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-282 of human SNRPA (NP_004587.1).
Purification Method Affinity purification
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6626
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze / thaw cycles.
Form Liquid
Isotype IgG
Research Category Other
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.