Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNRPB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584769
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584769 Supplier Invitrogen™ Supplier No. PA584769
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QAPAGLAGPV RGVGGPSQQV MTPQGRGT Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

The protein encoded by this gene is one of several nuclear proteins that are found in common among U1, U2, U4/U6, and U5 small ribonucleoprotein particles (snRNPs). These snRNPs are involved in pre-mRNA splicing, and the encoded protein may also play a role in pre-mRNA splicing or snRNP structure. Autoantibodies from patients with systemic lupus erythematosus frequently recognize epitopes on the encoded protein. Two transcript variants encoding different isoforms (B and B') have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRPB
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNRPB
Gene Accession No. P14678
Gene Alias AL024368; AU018828; B polypeptide of Sm protein; CCMS; COD; sm protein B; Sm protein B/B'; SM11; small nuclear ribonucleoprotein B; small nuclear ribonucleoprotein polypeptide B; small nuclear ribonucleoprotein polypeptides B and B'; small nuclear ribonucleoprotein polypeptides B and B1; small nuclear ribonucleoprotein-associated protein B; Small nuclear ribonucleoprotein-associated proteins B and B'; smB; sm-B; Sm-B/B'; SmB/B'; sm-B/Sm-B'; SmB/SmB'; SNRNP-B; Snrpb; SNRPB1
Gene Symbols SNRPB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNRPB. Recombinant protein control fragment (Product #RP-105439).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6628
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.