Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNRPD2 Antibody, mFluor Violet 610 SE, R&D Systems™
SDP

Catalog No. NB330651
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB330651 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB330651 Supplier R&D Systems Supplier No. NBP297664MFV610

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

SNRPD2 Polyclonal specifically detects SNRPD2 in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).

Specifications

Antigen SNRPD2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias small nuclear ribonucleoprotein D2 polypeptide (16.5kD), small nuclear ribonucleoprotein D2 polypeptide 16.5kDa, small nuclear ribonucleoprotein Sm D2, Sm-D2SMD2, snRNP core protein D2, SNRPD1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SNRPD2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6633
Target Species Human
Content And Storage Store at 4C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.