Learn More
Invitrogen™ SNRPG Polyclonal Antibody

Description
Immunogen sequence: MSKAHPPELKK FMDKKLSLKL NGGR Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Specifications
Specifications
| Antigen | SNRPG |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SNRPG |
| Gene Accession No. | P62308 |
| Gene Alias | 2810024K17Rik; AL022803; PBSCG; Sm protein G; Small nuclear ribonucleoprotein G; small nuclear ribonucleoprotein polypeptide G; SMG; Sm-G; snRNP-G; Snrpg |
| Gene Symbols | SNRPG |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.