Learn More
ATCC SNU-5 Purified Genomic DNA, Quantitative (MET Amplification), 3 μg
Supplier:  ATCC CRL5973DQ
Encompass
Purified, quantified genomic DNA extracted from the authenticated SNU-5 human gastric carcinoma cell line. Prepared as a research-grade control material and quantified for MET amplification assays using ddPCR.
  • Purified genomic DNA extracted from SNU-5 cell line.
  • Quantified for MET amplification by ddPCR.
  • Amount: 3 μg total DNA mass.
  • Supplied in 1X TE buffer.
  • Store at -20°C or colder.
  • Spin down before use.
  • For research use only; not for therapeutic, diagnostic, or consumption.
Catalog No. NC3253227