Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. APPH36 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APPH36 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APPH36 Supplier Invitrogen™ Supplier No. PA1836
  • $559.00
    $428.65 / Each of 1

    Save $130.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-836 detects SNX1 in human and mouse samples. It is recommended for use with recombinant protein. PA1-836 has been successfully used in Western blot and ICC/IF procedures. By Western blot, this antibody detects a ~60 kDa protein representing SNX1in human samples. The PA1-836 immunogen is a synthetic peptide corresponding to residues Q(139) E D Q F D L T V G I T D P E K I(155) of human SNX1. This peptide (Cat. # PEP-286) is available for use in neutralization and control experiments.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This endosomal protein regulates the cell-surface expression of epidermal growth factor receptor. This protein also has a role in sorting protease-activated receptor-1 from early endosomes to lysosomes. This protein may form oligomeric complexes with family members. This gene results in three transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene SNX1
Gene Accession No. Q13596, Q9WV80
Gene Alias HsT17379; MGC8664; Snx1; SNX1A; sorting nexin 1; sorting nexin 1A; sorting nexin-1; VPS5
Gene Symbols SNX1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues Q(139) E D Q F D L T V G I T D P E K I(155) of human SNX1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56440, 6642
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.