Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX10 Monoclonal Antibody (OTI6B7)
GREENER_CHOICE

Catalog No. PIMA527067
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527067 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527067 Supplier Invitrogen™ Supplier No. MA527067
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SNX10 Monoclonal Antibody for Western Blot, IHC (P)

Probable phosphoinositide-binding protein involved in protein sorting and membrane trafficking in endosomes. Plays a role in cilium biogenesis through regulation of the transport and the localization of proteins to the cilium. Required for the localization to the cilium of V-ATPase subunit ATP6V1D and ATP6V0D1, and RAB8A. Involved in osteoclast differentiation and therefore bone resorption.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SNX10
Gene Accession No. Q9Y5X0
Gene Alias 2410004M09Rik; OPTB8; Snx10; sorting nexin 10; sorting nexin-10; Sorting nexin-10-like protein
Gene Symbols SNX10
Host Species Mouse
Immunogen Full length human recombinant protein of SNX10 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29887
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.