Learn More
Essendant Antimicrobial Foam Skin Cleanser, Fresh Scent, 1,000 mL Bottle, 6/Carton
Supplier:  Essendant KCC91554CT
Encompass_Preferred
Kills harmful germs while moisturizing the skin. This formulation does not contain triclosan. Contains (0.1% Benzalkonium Chloride) to kill harmful germs such as E. Coli, Listeria and Salmonella. Contains aloe and glycerin for a smooth and creamy feel and for moisturizing. Formulation is mild and designed for everyday use.
Catalog No. 26-799-02
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.