Learn More
Teknova SOB Medium with Animal-Free Soytone. 1000mL, Sterile. 8 pack.
SDP
Supplier:  Teknova S26158PK
Encompass_Preferred
2.0% Animal-Free Soytone, 0.5% Yeast Extract, 10mM Sodium Chloride, 2.5mM Potassium Chloride, 10mM Magnesium Chloride, 10mM Magnesium Sulfate
Catalog No. 50-238-7699
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.