Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SOC (Powder)

Catalog No. MP113031012 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
227 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113031012 227 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP113031012 Supplier MP Biomedicals™ Supplier No. 113031012
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maximizes the transformation efficiency of competent cells. MP Biomedicals SOC (Powder) is a special medium for incubating competent cells in the recovery step of E. coli cell transformations. It allows expression of transferred resistance genes before ex

Chosen reagents are combined in the accurate proportions according to the formula and thoroughly blended for a uniform distribution. A sample of the blended formulation is used to prepare liquid. The combinations are mixed to dissolve components and then autoclaved at 121°C for 15 minutes.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified™
Content And Storage Store at room temperature.
Description SOC
Form Powder
Formulation Per liter: 20 g Tryptone-B; 5 g Yeast Extract-B; 0.5 g NaCl; 5 g MgSO4-7H2O; 20 mM glucose. Small quantities of biological buffers have been added for proper pH adjustment.
For Use With (Application) Recovery step of Escherichia coli competent cell transformations
Quantity 227 g
Regulatory Status RUO
Specimen Type E. coli
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.