Learn More
Grainger Socket Head Cap Screws
Supplier:  Grainger 22UD07
Encompass_Preferred
The Grainger website offers a wide selection of socket head cap screws in various sizes, materials, and finishes. These fasteners feature a cylindrical head with an internal hex drive for tightening with an Allen wrench. They are available in alloy steel, stainless steel, and other materials. Diameters range from #4 to 1 inch and lengths from 1/4 inch to 6 inches. Drive sizes include hex, 12-point, and 6-lobe options. Socket head cap screws can be used in many applications that require a strong, low-profile fastener. Grainger provides technical specs for each screw as well as convenient sorting and filtering options. Bulk quantities are available for many items.
Catalog No. NC2596497
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.