Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Socorex Qualitix™ 10 μL Micropipette Tips from DWK Life Sciences™
SDP

Catalog No. 12141503 Shop All DWK Life Sciences Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Format:
Bagged
Rack
Packaging:
1000/Bag
12 x 96/Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Filtered Format Packaging
12-141-503 Non-sterile Non-filtered Bagged 1000/Bag
12-141-504 Sterile Filtered Rack 12 x 96/Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-141-503 Supplier Socorex Supplier No. 3090.0010B
Add to Cart
Add to Cart

Certified free of nuclease, endotoxin and ATP

  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Length (Metric) 31.4 mm
Certifications/Compliance Nuclease-free (DNase and RNase), Human DNA-free (bioproof purity level), Pyrogen-free, ATP-free (Adenosin Triphosphate), Heavy metal-free, Protease-free
Clean Claims RNase, DNase, and pyrogen free
Format Bagged
Tip Style Fine Point
Packaging 1000/Bag
Product Type Micropipette tips
For Use With (Equipment) Universal Fit Micro
Graduated Yes
Product Line Socorex Qualitix
Material Virgin Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.