Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Socorex Qualitix™ 10 μL Micropipette Tips Low Binding from DWK Life Sciences™
SDP

Catalog No. 12141525 Shop All DWK Life Sciences Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Filtered
12-141-525 Sterile Non-filtered
12-141-524 Non-sterile Filtered
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-141-525 Supplier Socorex Supplier No. 3090.0010PRS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Certified free of nuclease, endotoxin and ATP

  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Autoclavable Autoclavable
Length (Metric) 31.4 mm
Certifications/Compliance Certified free of nuclease, endotoxin, and ATP. Free of detectable DNase, RNase, and Pyrogen. Certified free of human DNA and protease. Gamma irradiation certified. Filtered.
Clean Claims RNase, DNase, and pyrogen free
Format Rack
Tip Style Low Binding
Packaging 12 x 96/Rack
Quantity 12 x 96/Rack
Product Type Micropipette tips
For Use With (Equipment) Universal Fit Micro
Graduated Yes
Product Line Socorex Qualitix
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.