Learn More
Socorex Qualitix™ 1000 μL Micropipette Tips Low Binding from DWK Life Sciences™
Shop All DWK Life Sciences Products

Description
- Wide selection, superior quality, autoclavable
- Bags, racks and tipfill rack filling system
- Lot by lot consistency
- Reduced packaging size, less bench space requirement
- Broad compatibility with major micropipette brands
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Natural |
| Autoclavable | Autoclavable |
| Length (Metric) | 88.9 mm |
| Certifications/Compliance | Protease-free certified, Certified free of nuclease, endotoxin and ATP, Human DNA-free, Heavy metal-free |
| Clean Claims | RNase, DNase, and pyrogen free |
| For Use With (Application) | Proteomic research, accurate handling of viscous liquids, safe handling of samples containing genetic material (PCR, qPCR), cell culture, accurate measurement of cells or bacteria activity, trace analysis, protection of valuable samples containing peptides and proteins |
| Format | Rack |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.