Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Socorex Qualitix™ 1000 μL Micropipette Tips Rack Refill from DWK Life Sciences™
SDP

Catalog No. 12141522 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12-141-522 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-141-522 Supplier Socorex Supplier No. 3070.1000T
Add to Cart
Add to Cart

Bags, racks and tipfill rack filling system

  • Wide selection, superior quality, autoclavable
  • Lot by lot consistency
  • Certified free of nuclease, endotoxin and ATP
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Color Natural
Autoclavable Autoclavable
Length (Metric) 88.9 mm
Certifications/Compliance Free of nuclease, endotoxin and ATP
Clean Claims RNase, DNase, and pyrogen free
For Use With (Application) Broad compatibility with major micropipette brands
Packaging 5 x 96/Rack
Quantity 480/Pk. (5x 96/Rack)
Product Type Micropipette tips rack refill
Includes Rack
For Use With (Equipment) Universal Fit Micro
Product Line Qualitix
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.