Learn More
Socorex Qualitix™ 1000 μL Premium Micropipette Tips, Tip Refilling System from DWK Life Sciences™
SDP
Catalog No. 12141523 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
1000 μL
Catalog No. Size
12-141-523 1000 μL
1 options
Catalog No. 12-141-523 Supplier Socorex Supplier No. 3070.1000ER
Add to Cart
Add to Cart
Certified free of nuclease, endotoxin and ATP
  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
TRUSTED_SUSTAINABILITY
Specifications
Clean Claims DNase, RNase, and pyrogen free
Autoclavable Autoclavable
Color Natural
Size 1000 μL
Length (Metric) 49.6 mm