Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Socorex Qualitix™ 350 μL Micropipette Tips from DWK Life Sciences™
SDP

Catalog No. 12141516 Shop All DWK Life Sciences Products
Change view
Click to view available options
Format:
Bagged
Rack
Packaging:
10 x 96/Rack
1000/Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Packaging
12-141-516 Bagged 1000/Bag
12-141-517 Rack 10 x 96/Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-141-516 Supplier Socorex Supplier No. 308.0350B
Add to Cart
Add to Cart

Certified free of nuclease, endotoxin and ATP

  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 350 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Length (Metric) 58.8 mm
Certifications/Compliance ISO 8655/17025, ISO, SQS
Clean Claims RNase, DNase, and pyrogen free
For Use With (Application) Optimal tightness fitting on Socorex pipettes; Broad compatibility with other brands; Compatible with Acura manual and Acura electro pipettes in volume ranges: 120 to 200 μL, 10 to 100 μL, 20 to 200 μL, 40 to 350 μL
Format Bagged
Packaging 1000/Bag
Quantity 1000/Bag
Product Type Micropipette tips
For Use With (Equipment) Universal Fit Micro
Product Line Socorex Qualitix
Material Virgin Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.