Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Socorex Qualitix™ Micropipette Filter Tips from DWK Life Sciences™
SDP

Catalog No. 12141510 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
100 μL
200 μL
300 μL
1000 μL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12-141-510 100 μL
12-141-505 10 μL
12-141-509 20 μL
12-141-513 200 μL
12-141-518 300 μL
12-141-521 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 12-141-510 Supplier Socorex Supplier No. 3080.0100FRS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sterile Socorex™ Qualitix™ Micropipette Filter Tips are certified free of nuclease, endotoxin, and ATP.

  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
  • Sterile
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 μL
Volume (Metric) Dispensing 100 μL
Filtered Filtered
Sterility Sterile
Color Natural
Autoclavable Autoclavable
Length (Metric) 49.6 mm
Certifications/Compliance ISO 9001
Clean Claims RNase, DNase, and pyrogen free
For Use With (Application) Liquid Handling
Format Rack
Tip Style Beveled
Packaging 10 x 96/Rack
Quantity 960/Cs.
Product Type Micropipette Tips
Green Features Reduced Packaging Size
For Use With (Equipment) Universal Fit Micro
Product Line Qualitix
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.