Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Socorex Qualitix™ Micropipette Tips, Gel Loading from DWK Life Sciences™
SDP

Catalog No. 12141515 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
Length (Metric):
71 mm
82.5 mm
Packaging:
4 x 204/Rack
6 x 96/Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Length (Metric) Packaging
12-141-515 200 μL 82.5 mm 6 x 96/Rack
12-141-507 10 μL 71 mm 4 x 204/Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-141-515 Supplier Socorex Supplier No. 3080.0200GR
Add to Cart
Add to Cart

Certified free of nuclease, endotoxin and ATP

  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Length (Metric) 82.5 mm
Certifications/Compliance ISO 11137
Clean Claims RNase, DNase, and pyrogen free
For Use With (Application) Precise Liquid Distribution
Format Rack
Tip Style Gel Loading
Packaging 6 x 96/Rack
Product Type Micropipette tips, Gel loading
For Use With (Equipment) Universal Fit Micro
Graduated No
Product Line Socorex Qualitix
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.