Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOCS3 Monoclonal Antibody (SO1)
GREENER_CHOICE

Catalog No. 01676129 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-129 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-129 Supplier Invitrogen™ Supplier No. MA119373
  • $671.00
    $550.65 / Each of 1

    Save $120.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This antibody reacts with SOCS3, an intracellular cytokine signaling inhibitor. Western Blot: Reducing conditions.

Accumulating evidence demonstrates that cytokine receptor signaling is negatively regulated by a family of Src homology 2 domain-containing adaptor molecules Termed SOCS (Suppressor of Cytokine Signaling). To date, there are eight members of SOCS family have been recognized, they are SOCS-1, 2, 3, 4, 5, 6, 7 and CIS. Structurally the SOCS proteins are composed of an N-terminal region of variable length and amino acid composition, a central SH2 domain, and a previously unrecognized C-terminal motif that we have called the SOCS box. The SOCS proteins appear to form part of a classical negative feed back loop that regulates cytokine signal transduction via a STAT-induced transcriptional mechanism. Transcription of each of the SOCS genes occurs rapidly in vitro and in vivo in response to cytokines, and once produced, the various members of the SOCS family appear to inhibit signaling in different ways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOCS3
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone SO1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene Socs3
Gene Accession No. O14543
Gene Alias ATOD4; CIS3; CIS-3; Cish3; cytokine inducible SH2-containing protein 3; cytokine-inducible SH2 protein 3; E2a-Pbx1 target gene in fibroblasts 10; Ef10; EF-10; MGC71791; Protein EF-10; Socs3; Socs-3; SSI3; SSI-3; STAT-induced STAT inhibitor 3; Suppressor of cytokine signaling 3
Gene Symbols Socs3
Host Species Mouse
Immunogen Full-length SOCS3 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9021
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.