Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More

Ethylenediaminetetraacetic Acid Disodium Salt, Standard Solution (0.1 M/Certified), Fisher Chemical™ PROMO
Synonym: EDTA Disodium Salt, Edetate Disodium, Disodium Edetate
Quantity | 1 L |
---|
Description

Chemical Identifiers
6381-92-6, 7732-18-5 | |
372.24 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
44120005 | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
C10H18N2Na2O10 | |
MFCD00150037,MFCD00003541 | |
edta disodium salt | |
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |
Specifications
Colorless | |
100°C | |
C10H18N2Na2O10 | |
0.0995 to 0.1005 M | |
edta disodium salt | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
372.24 | |
Certified | |
14 mmHg | |
1 L | |
Ethylenediamine Tetraacetic Acid, Disodium Salt, Standard Solution |
6381-92-6, 7732-18-5 | |
0°C | |
MFCD00150037,MFCD00003541 | |
1.02 g/cm3 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
44120005 | |
Poly Bottle | |
4.4 to 4.5 | |
Liquid |
Safety and Handling

Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction