Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-decanesulfonate, 99%

Catalog No. AAA1463806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1463806 5 g
AAA1463814 25 g
AAA1463822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1463806 Supplier Thermo Scientific Chemicals Supplier No. A1463806
Add to Cart

CAS: 13419-61-9 | C10H21NaO3S | 244.325 g/mol

Ion-associating reagent for HPLC, useful for analysis of peptides and proteins.Sodium 1-decanesulfonate is used as a base in coupling reactions such as Suzuki coupling. It is used as an ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is used to study the mechanism of guanidine compounds on a porous graphite carbon and electro reduction of Zn2+ ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ion-associating reagent for HPLC, useful for analysis of peptides and proteins.Sodium 1-decanesulfonate is used as a base in coupling reactions such as Suzuki coupling. It is used as an ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is used to study the mechanism of guanidine compounds on a porous graphite carbon and electro reduction of Zn2+ ions.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13419-61-9
Molecular Formula C10H21NaO3S
Molecular Weight (g/mol) 244.325
MDL Number MFCD00007526
InChI Key AIMUHNZKNFEZSN-UHFFFAOYSA-M
Synonym sodium decane-1-sulfonate, sodium 1-decanesulfonate, 1-decanesulfonic acid sodium salt, 1-decanesulfonic acid, sodium salt, decyl sodium sulfonate, sodium decane-1-sulphonate, ipc-alks-10, n-decylsulfonic acid, sodium salt, 1-decanesulphonic acid, sodium salt, 2-decanesulfonic acid, sodium salt
PubChem CID 2724181
IUPAC Name sodium;decane-1-sulfonate
SMILES CCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Assay Percent Range 99% (dry wt.), water <1.5%
Quantity 5 g
Beilstein 3918920
Solubility Information Soluble in water.
Formula Weight 244.33
Percent Purity 99%
Assay (dry wt.), water <1.5%
Chemical Name or Material Sodium 1-decanesulfonate
EINECSNumber 236-525-9
TSCA No
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.