Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sodium 1-dodecanesulfonate, 98%

Catalog No. AAH3659009
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH36590-09 10 g
AAH36590-03 1 g
AAH36590-14 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH36590-09 Supplier Thermo Scientific™ Supplier No. H3659009
Add to Cart

Chemical Identifiers

CAS 2386-53-0
Molecular Formula C12H25NaO3S
Molecular Weight (g/mol) 272.38
MDL Number MFCD00007527
InChI Key DAJSVUQLFFJUSX-UHFFFAOYSA-M
Synonym 1-Dodecanesulfonic acid sodium salt;
IUPAC Name sodium dodecane-1-sulfonate
SMILES [Na+].CCCCCCCCCCCCS([O-])(=O)=O

Specifications

Melting Point >300°
Physical Form Solid
Quantity 10 g
Linear Formula CH{3}(CH{2}){1}{1}SO{3}Na
Formula Weight 272.39
Percent Purity 98%
ShelfLife 5 Years
EINECSNumber 219-200-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.