Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-heptanesulfonate monohydrate, 99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111013498 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3498 5 g
11-101-3496 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3498 Supplier MilliporeSigma Supelco Supplier No. 518335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium 1-heptanesulfonate monohydrate can be used as an ion-pairing (IP) reagent, to develop ion-pair reversed phase liquid chromatography method, for the determination of antituberculosis drug ethambutol hydrochloride.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207300-90-1
Molecular Formula C7H17NaO4S
Molecular Weight (g/mol) 220.26
MDL Number MFCD00149550
InChI Key XWZCREJRXRKIRQ-UHFFFAOYSA-M
Synonym 1-Heptanesulfonic acid sodium salt monohydrate
IUPAC Name sodium heptane-1-sulfonate hydrate
SMILES O.[Na+].CCCCCCCS([O-])(=O)=O

Specifications

Linear Formula CH3(CH2)5 CH2SO3Na · H2O
Quantity 5 g
Formula Weight 220.26
Percent Purity ≥99.0% (T)
Grade For ion pair chromatography
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.