Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-nonanesulfonate, ≥99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111014002 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4002 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4002 Supplier MilliporeSigma Supelco Supplier No. 7431610GF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00025010
Synonym 1-Nonanesulfonic acid sodium salt

Specifications

Melting Point >300°C
CAS 35192-74-6
Packaging Glass Bottle
Linear Formula CH3(CH2)8 SO3 Na
Quantity 10 g
Formula Weight 230.30
Percent Purity ≥99.0% (T)
Grade For ion pair chromatography
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.