Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-Octanesulfonate 98.0+%, TCI America™
SDP

Catalog No. I034625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I034625G 25 g
I03465G 5 g
I0346100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I034625G Supplier TCI America Supplier No. I034625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium 1-Octanesulfonate [Reagent for Ion-Pair Chromatography], >-98.0% (T), C8H17NaO3S, CAS Number-5324-84-5

Sodium 1-Octanesulfonate [Reagent for Ion-Pair Chromatography], >-98.0% (T), C8H17NaO3S, CAS Number-5324-84-5

Chemical Identifiers

CAS 5324-84-5
Molecular Formula C8H17NaO3S
Molecular Weight (g/mol) 216.27
MDL Number MFCD00007544
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
Synonym sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
PubChem CID 23669624
IUPAC Name sodium octane-1-sulfonate
SMILES [Na+].CCCCCCCCS([O-])(=O)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 216.27
Percent Purity ≥98.0% (T)
Chemical Name or Material Sodium 1-Octanesulfonate [Reagent for Ion-Pair Chromatography]
EINECSNumber (2)-1640
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.