CAS RN 5324-84-5
1-Octanesulfonic Acid, Sodium Salt Anhydrous, HPLC Grade, Thermo Scientific Chemicals
CAS: 5324-84-5 Molecular Formula: C8H17NaO3S Molecular Weight (g/mol): 216.27 MDL Number: MFCD00007544 InChI Key: HRQDCDQDOPSGBR-UHFFFAOYSA-M Synonym: sodium 1-octanesulfonate,sodium octane-1-sulfonate,1-octanesulfonic acid sodium salt,sodium octanesulfonate,1-octanesulfonic acid, sodium salt,1-octanesulfonate, sodium,sodium octylsulfonate,unii-j46ez5jyb3,octyl sodium sulfonate,ipc-alks-8 PubChem CID: 23669624 IUPAC Name: sodium octane-1-sulfonate SMILES: [Na+].CCCCCCCCS([O-])(=O)=O
1 Octane Sulfonic Acid Sodium Salt, HPLC Ion Pair Agent, Fisher Chemical™
CAS: 5324-84-5 Molecular Formula: C8H17NaO3S Molecular Weight (g/mol): 216.27 MDL Number: MFCD00007544 InChI Key: HRQDCDQDOPSGBR-UHFFFAOYSA-M IUPAC Name: sodium octane-1-sulfonate SMILES: [Na+].CCCCCCCCS([O-])(=O)=O
Sodium 1-octanesulfonate, 99%
CAS: 5324-84-5 Molecular Formula: C8H17NaO3S Molecular Weight (g/mol): 216.27 MDL Number: MFCD00007544 InChI Key: HRQDCDQDOPSGBR-UHFFFAOYSA-M Synonym: sodium 1-octanesulfonate,sodium octane-1-sulfonate,1-octanesulfonic acid sodium salt,sodium octanesulfonate,1-octanesulfonic acid, sodium salt,1-octanesulfonate, sodium,sodium octylsulfonate,unii-j46ez5jyb3,octyl sodium sulfonate,ipc-alks-8 PubChem CID: 23669624 IUPAC Name: sodium;octane-1-sulfonate SMILES: [Na+].CCCCCCCCS([O-])(=O)=O
1-Octanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
CAS: 5324-84-5 Molecular Formula: C8H17NaO3S Molecular Weight (g/mol): 216.27 MDL Number: MFCD00007544 InChI Key: HRQDCDQDOPSGBR-UHFFFAOYSA-M Synonym: sodium 1-octanesulfonate,sodium octane-1-sulfonate,1-octanesulfonic acid sodium salt,sodium octanesulfonate,1-octanesulfonic acid, sodium salt,1-octanesulfonate, sodium,sodium octylsulfonate,unii-j46ez5jyb3,octyl sodium sulfonate,ipc-alks-8 PubChem CID: 23669624 IUPAC Name: sodium;octane-1-sulfonate SMILES: [Na+].CCCCCCCCS([O-])(=O)=O
1-Octanesulfonic acid, sodium salt, HPLC grade
CAS: 5324-84-5 Molecular Formula: C8H17NaO3S Molecular Weight (g/mol): 216.27 MDL Number: MFCD00007544 InChI Key: HRQDCDQDOPSGBR-UHFFFAOYSA-M Synonym: sodium 1-octanesulfonate,sodium octane-1-sulfonate,1-octanesulfonic acid sodium salt,sodium octanesulfonate,1-octanesulfonic acid, sodium salt,1-octanesulfonate, sodium,sodium octylsulfonate,unii-j46ez5jyb3,octyl sodium sulfonate,ipc-alks-8 PubChem CID: 23669624 SMILES: [Na+].CCCCCCCCS([O-])(=O)=O
LiChropur™ Sodium 1-octanesulfonate monohydrate , ≥99.0% (T), MilliporeSigma™ Supelco™
MDL Number: MFCD00149551 Synonym: 1-Octanesulfonic acid sodium salt monohydrate
1-Octanesulfonic Acid Sodium Salt, HPLC Grade, J.T. Baker™
CAS: 5324-84-5 Molecular Formula: C8H17NaO3S Molecular Weight (g/mol): 216.27 MDL Number: MFCD00007544 InChI Key: HRQDCDQDOPSGBR-UHFFFAOYSA-M Synonym: sodium 1-octanesulfonate,sodium octane-1-sulfonate,1-octanesulfonic acid sodium salt,sodium octanesulfonate,1-octanesulfonic acid, sodium salt,1-octanesulfonate, sodium,sodium octylsulfonate,unii-j46ez5jyb3,octyl sodium sulfonate,ipc-alks-8 PubChem CID: 23669624 SMILES: [Na+].CCCCCCCCS([O-])(=O)=O
Octane-1-sulfonic acid sodium salt, LiChropur™, MilliporeSigma™
CAS: 5324-84-5 Molecular Formula: C8H17NaO3S Molecular Weight (g/mol): 216.27 MDL Number: MFCD00007544 InChI Key: HRQDCDQDOPSGBR-UHFFFAOYSA-M Synonym: sodium 1-octanesulfonate,sodium octane-1-sulfonate,1-octanesulfonic acid sodium salt,sodium octanesulfonate,1-octanesulfonic acid, sodium salt,1-octanesulfonate, sodium,sodium octylsulfonate,unii-j46ez5jyb3,octyl sodium sulfonate,ipc-alks-8 PubChem CID: 23669624 IUPAC Name: sodium octane-1-sulfonate SMILES: [Na+].CCCCCCCCS([O-])(=O)=O
Sodium 1-Octanesulfonate, 98+ Percent, Reagent, Spectrum™ Chemical
CAS: 5324-84-5
1-Octanesulfonic Acid, Sodium Salt OR, Macron Fine Chemicals™
CAS: 5324-84-5 Molecular Formula: C8H17NaO3S Molecular Weight (g/mol): 216.27 MDL Number: MFCD00007544 InChI Key: HRQDCDQDOPSGBR-UHFFFAOYSA-M Synonym: sodium 1-octanesulfonate,sodium octane-1-sulfonate,1-octanesulfonic acid sodium salt,sodium octanesulfonate,1-octanesulfonic acid, sodium salt,1-octanesulfonate, sodium,sodium octylsulfonate,unii-j46ez5jyb3,octyl sodium sulfonate,ipc-alks-8 PubChem CID: 23669624 IUPAC Name: sodium octane-1-sulfonate SMILES: [Na+].CCCCCCCCS([O-])(=O)=O
Octane-1-sulfonic acid sodium salt, LiChropur™ EMD-Millipore
CAS: 5324-84-5 Molecular Formula: C8H17NaO3S Molecular Weight (g/mol): 216.27 MDL Number: MFCD00007544 InChI Key: HRQDCDQDOPSGBR-UHFFFAOYSA-M Synonym: sodium 1-octanesulfonate,sodium octane-1-sulfonate,1-octanesulfonic acid sodium salt,sodium octanesulfonate,1-octanesulfonic acid, sodium salt,1-octanesulfonate, sodium,sodium octylsulfonate,unii-j46ez5jyb3,octyl sodium sulfonate,ipc-alks-8 PubChem CID: 23669624 IUPAC Name: sodium octane-1-sulfonate SMILES: [Na+].CCCCCCCCS([O-])(=O)=O
Sodium 1-Octanesulfonate 98.0+%, TCI America™
CAS: 5324-84-5 Molecular Formula: C8H17NaO3S Molecular Weight (g/mol): 216.27 MDL Number: MFCD00007544 InChI Key: HRQDCDQDOPSGBR-UHFFFAOYSA-M Synonym: sodium 1-octanesulfonate,sodium octane-1-sulfonate,1-octanesulfonic acid sodium salt,sodium octanesulfonate,1-octanesulfonic acid, sodium salt,1-octanesulfonate, sodium,sodium octylsulfonate,unii-j46ez5jyb3,octyl sodium sulfonate,ipc-alks-8 PubChem CID: 23669624 IUPAC Name: sodium octane-1-sulfonate SMILES: [Na+].CCCCCCCCS([O-])(=O)=O
Sodium 1-Octanesulfonate 98.0+%, TCI America™
CAS: 5324-84-5 Molecular Formula: C8H17NaO3S Molecular Weight (g/mol): 216.27 MDL Number: MFCD00007544 InChI Key: HRQDCDQDOPSGBR-UHFFFAOYSA-M Synonym: sodium 1-octanesulfonate,sodium octane-1-sulfonate,1-octanesulfonic acid sodium salt,sodium octanesulfonate,1-octanesulfonic acid, sodium salt,1-octanesulfonate, sodium,sodium octylsulfonate,unii-j46ez5jyb3,octyl sodium sulfonate,ipc-alks-8 PubChem CID: 23669624 IUPAC Name: sodium octane-1-sulfonate SMILES: [Na+].CCCCCCCCS([O-])(=O)=O
Sodium 1-octanesulfonate hydrate, HPLC grade, 99+%
CAS: 5324-84-5 Molecular Formula: C8H17NaO3S Molecular Weight (g/mol): 216.27 MDL Number: MFCD00007544 InChI Key: HRQDCDQDOPSGBR-UHFFFAOYSA-M Synonym: sodium 1-octanesulfonate,sodium octane-1-sulfonate,1-octanesulfonic acid sodium salt,sodium octanesulfonate,1-octanesulfonic acid, sodium salt,1-octanesulfonate, sodium,sodium octylsulfonate,unii-j46ez5jyb3,octyl sodium sulfonate,ipc-alks-8 PubChem CID: 23669624 IUPAC Name: sodium;octane-1-sulfonate SMILES: [Na+].CCCCCCCCS([O-])(=O)=O