Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-Tetradecanesulfonate, TCI America™
SDP

Catalog No. T07571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T07571G Supplier TCI America Supplier No. T07571G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6994-45-2
Molecular Formula C14H29NaO3S
Molecular Weight (g/mol) 300.433
MDL Number MFCD00043415
InChI Key AYFACLKQYVTXNS-UHFFFAOYSA-M
Synonym 1-Tetradecanesulfonic Acid Sodium Salt, Tetradecylsulfonic Acid Sodium Salt
PubChem CID 23693105
IUPAC Name sodium;tetradecane-1-sulfonate
SMILES CCCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 227°C
Color White
Quantity 1 g
Formula Weight 300.43
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Tetradecanesulfonate
EINECSNumber (2)-1640
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.