Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-Undecanesulfonate 98.0+%, TCI America™
SDP

Catalog No. U003125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U003125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U003125G Supplier TCI America Supplier No. U003125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5838-34-6
Molecular Formula C20H14N4O4S
Molecular Weight (g/mol) 406.42
MDL Number MFCD00053627
InChI Key RFNALTSHRWGOHV-UHFFFAOYSA-N
Synonym sodium 1-undecanesulfonate, sodium undecane-1-sulfonate, 1-undecanesulfonic acid sodium salt, sodium undecylsulfonate, ipc-alks-11, acmc-1bn99, 1-undecanesulfonicacid, sodium salt 1:1
PubChem CID 23692564
IUPAC Name 4-[2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)acetamido]-N-(1,3-thiazol-2-yl)benzamide
SMILES O=C(CN1C(=O)C2=CC=CC=C2C1=O)NC1=CC=C(C=C1)C(=O)NC1=NC=CS1

Specifications

Color White
Quantity 25 g
Formula Weight 258.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Undecanesulfonate
EINECSNumber (2)-1640
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.