Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 2,6-Dinitro-4-(trifluoromethyl)benzenesulfonate 95.0+%, TCI America™
SDP

Catalog No. D201125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D201125G 25 g
D20115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D201125G Supplier TCI America Supplier No. D201125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54495-25-9
Molecular Formula C7H2F3N2NaO7S
Molecular Weight (g/mol) 338.145
MDL Number MFCD00059068
InChI Key PYGGEZQHHQFWCY-UHFFFAOYSA-M
Synonym 2,6-Dinitro-4-(trifluoromethyl)benzenesulfonic Acid Sodium Salt
PubChem CID 2737132
IUPAC Name sodium;2,6-dinitro-4-(trifluoromethyl)benzenesulfonate
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])S(=O)(=O)[O-])[N+](=O)[O-])C(F)(F)F.[Na+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 338.14
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2,6-Dinitro-4-(trifluoromethyl)benzenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.