Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 2-chloroethanesulfonate hydrate, 98+% (dry wt.), water <10%

Catalog No. AAA1999514
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1999514 25 g
AAA1999506 5 g
AAA1999522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1999514 Supplier Thermo Scientific Chemicals Supplier No. A1999514
Add to Cart

CAS: 15484-44-3 | C2H4ClNaO3S | 166.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15484-44-3
Molecular Formula C2H4ClNaO3S
Molecular Weight (g/mol) 166.55
MDL Number MFCD00149546 MFCD00007531
InChI Key BVIXLMYIFZGRBH-UHFFFAOYSA-M
Synonym sodium 2-chloroethylsulfonate, 2-chloroethane sulfonic acid sodium salt, sodium .beta.-chloroethanesulfonate, sodium 2-chloroethane-1-sulfonic acid, sodium 2-chloroethanesulfonic acid
PubChem CID 54611855
SMILES [Na+].[O-]S(=O)(=O)CCCl

Specifications

Melting Point ∼292°C (decomposition)
Quantity 25 g
Beilstein 5649202
Sensitivity Hygroscopic
IUPAC Name 2-chloroethane-1-sulfonic acid sodium
Formula Weight 166.56 (Anhydrous)
Percent Purity ≥98%
Assay (dry wt.), water <10%
Chemical Name or Material Sodium 2-chloroethanesulfonate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-508-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.