Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 2-mercaptoethanesulfonate, Analytical Standard, 98.0% (titration), MilliporeSigma™ Supelco™

Catalog No. 111013438 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3438 25 g
11-101-3440 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3438 Supplier MilliporeSigma Supelco Supplier No. M151125G

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

Specifications

Packaging Poly Bottle
CAS 19767-45-4
Physical Form Neat
Linear Formula HSCH2CH2SO3Na
MDL Number MFCD00007535
Synonym 2-Mercaptoethanesulfonic acid sodium salt; Coenzyme M sodium salt; HS-CoM Na; MESNA
Quantity 25 g
Formula Weight 164.18
Percent Purity ≥98.0% (titration)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.