Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 2-Nitroaniline-4-sulfonate 98.0+%, TCI America™
SDP

Catalog No. N037125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N037125G 25 g
N0371100G 100 g
N0371500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N037125G Supplier TCI America Supplier No. N037125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5042-33-1
Molecular Formula C6H5N2NaO5S
Molecular Weight (g/mol) 240.165
MDL Number MFCD00070599
InChI Key SXTOGAIYFMTUAD-UHFFFAOYSA-M
Synonym 4-Amino-3-nitrobenzenesulfonic Acid Sodium Salt, 2-Nitroaniline-4-sulfonic Acid Sodium Salt, Sodium 4-Amino-3-nitrobenzenesulfonate
PubChem CID 165281
IUPAC Name sodium;4-amino-3-nitrobenzenesulfonate
SMILES C1=CC(=C(C=C1S(=O)(=O)[O-])[N+](=O)[O-])N.[Na+]

Specifications

Quantity 25 g
Formula Weight 240.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Nitroaniline-4-sulfonate
EINECSNumber (3)-2079
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.