Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 3-Chloro-2-hydroxypropanesulfonate 95.0+%, TCI America™
SDP

Catalog No. C1144500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1144500G 500 g
C114425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1144500G Supplier TCI America Supplier No. C1144500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 126-83-0
Molecular Formula C3H6ClNaO4S
Molecular Weight (g/mol) 196.577
MDL Number MFCD00013378
InChI Key TZLNJNUWVOGZJU-UHFFFAOYSA-M
Synonym 3-Chloro-2-hydroxypropanesulfonic Acid Sodium Salt
PubChem CID 23662382
IUPAC Name sodium;3-chloro-2-hydroxypropane-1-sulfonate
SMILES C(C(CCl)O)S(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 500 g
Formula Weight 196.58
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-Chloro-2-hydroxypropanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.