Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 3-(N-Ethylanilino)propanesulfonate 98.0+%, TCI America™
SDP

Catalog No. S08171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08171G 1 g
S0817200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08171G Supplier TCI America Supplier No. S08171G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82611-85-6
Molecular Formula C11H16NNaO3S
Molecular Weight (g/mol) 265.303
MDL Number MFCD01311047
InChI Key FFJBIXKLISICDT-UHFFFAOYSA-M
Synonym 3-(N-Ethylanilino)propanesulfonic Acid Sodium Salt, N-Ethyl-N-(3-sulfopropyl)aniline Sodium Salt, ALPS
PubChem CID 23685294
IUPAC Name sodium;3-(N-ethylanilino)propane-1-sulfonate
SMILES CCN(CCCS(=O)(=O)[O-])C1=CC=CC=C1.[Na+]

Specifications

Color White
Quantity 1 g
Formula Weight 265.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-(N-Ethylanilino)propanesulfonate [for Biochemical Research]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.