Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 3-Nitrosalicylate 99.0+%, TCI America™
SDP

Catalog No. N025825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N025825G 25 g
N02585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N025825G Supplier TCI America Supplier No. N025825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 164915-53-1
Molecular Formula C7H4NNaO5
Molecular Weight (g/mol) 205.10
MDL Number MFCD00070529
InChI Key DLXOJVOMIITILS-UHFFFAOYSA-M
Synonym 3-Nitrosalicylic Acid Sodium Salt
PubChem CID 54726778
IUPAC Name sodium 2-carboxy-6-nitrobenzen-1-olate
SMILES [Na+].OC(=O)C1=C([O-])C(=CC=C1)[N+]([O-])=O

Specifications

Quantity 25 g
Formula Weight 205.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-Nitrosalicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.