Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium 35 dichloro 2 hydro100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D4645
Encompass
Sodium 3 5-dichloro-2-hydroxybenzenesulfonate is used in conjunction with 4-aminoantipyrine (4-AAP) and hydrogen peroxide (H2O2) for chromogenic quantitation of uric acid in serum and urine samples.
Catalog No. 50-290-7267
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.