Learn More
Selleck Chemical LLC Sodium 4-Aminosalicylate S4073-50mg
SDP
Supplier:  Selleck Chemical LLC S407350MG
Encompass_Preferred
Sodium 4-Aminosalicylate is an antibiotic used to treat tuberculosis via NF- B inhibition and free radical scavenging
Catalog No. 50-310-9162
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.