Learn More
Sigma Organic Chemistry Sodium 4-ethynylbenzoat | 250mg | MFCD03426026
Supplier: Sigma Organic Chemistry CDS000957250MG

Sodium 4-ethynylbenzoat , 250mg
About this Item:
MDL #: MFCD03426026
Molecular Weight: 168.12
UNSPSC Code: 12352200
Chemical Formula: C9H5NaO2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.