Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 4-n-Octylbenzenesulfonate 98.0+%, TCI America™
SDP

Catalog No. O02035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02035G 5 g
O020325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02035G Supplier TCI America Supplier No. O02035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6149-03-7
Molecular Formula C14H21NaO3S
Molecular Weight (g/mol) 292.37
MDL Number MFCD00007512
InChI Key ASJUTVUXOMPOQH-UHFFFAOYSA-M
Synonym sodium 4-n-octylbenzenesulfonate, sodium 4-octylbenzenesulfonate, unii-hts5em67p9, hts5em67p9, 4-octylbenzenesulfonic acid sodium salt, 4-n-octylbenzenesulfonic acid sodium salt, acmc-2097it, dsstox_cid_27594, dsstox_rid_82441, dsstox_gsid_47594
PubChem CID 23670838
IUPAC Name sodium;4-octylbenzenesulfonate
SMILES CCCCCCCCC1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 300°C
Color White
Quantity 5 g
Formula Weight 292.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-n-Octylbenzenesulfonate
EINECSNumber (3)-1884&(3)-1906
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.