Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Abietate 95.0+%, TCI America™
SDP

Catalog No. A065125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A065125G 25 g
A06515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A065125G Supplier TCI America Supplier No. A065125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14351-66-7
Molecular Formula C20H29NaO2
Molecular Weight (g/mol) 324.44
MDL Number MFCD00058941
InChI Key ITCAUAYQCALGGV-XTICBAGASA-M
Synonym Abietic Acid Sodium Salt
PubChem CID 133121851
IUPAC Name sodium (1R,4aR,4bR,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylate
SMILES [Na+].CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C([O-])=O)[C@H]2CC1

Specifications

Quantity 25 g
Formula Weight 324.44
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Abietate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.