Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate, 0.100 Normal (N/10) in Glacial Acetic Acid, Reagents

Catalog No. 01333772
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-772 10 L
01-333-771 1 L
01-333-770 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-333-772 Supplier Reagents Holdings Llc Supplier No. CS12310010F
Add to Cart
Add to Cart

Sodium Acetate, 0.100 Normal (N/10) in Glacial Acetic Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 64-19-7 , 127-09-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Boiling Point 118°C
Color Colorless
pH <2
Physical Form Liquid
Quantity 10 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.0522
Chemical Name or Material Sodium Acetate, 0.100 Normal (N/10) in Glacial Acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.