Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium acetate, anhydrous, 99.997% (metals basis)

Catalog No. AA1100514
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1100514 25 g
AA1100506 5 g
AA1100522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1100514 Supplier Thermo Scientific Chemicals Supplier No. 01100514
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 127-09-3 ∣ C2H3NaO2 ∣ 82.03 g/mol

Sodium acetate is used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, as a plating agent, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography. It is used as a base in several organic reactions in research laboratories and industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Specifications

Melting Point 324°C
Color Black
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.997% (metals basis)
Beilstein 3595639
Solubility Information Very soluble in water. Soluble in alcohol.
Formula Weight 82.03
Percent Purity 100%
Grade Ungraded
Odor Nearly odorless
Sensitivity Hygroscopic
Appearance White crystalline powder
Density 1.528 g/cm3 at 20°C
Chemical Name or Material Sodium acetate, anhydrous
Show More Show Less
Hazard Category H500
EINECSNumber 204-823-8
RTECSNumber AJ4300010
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.