Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate Anhydrous ACS, Reagents

Catalog No. 01337769
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-769 100 g
01-337-767 3 kg
01-337-766 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-769 Supplier Reagents Holdings Llc Supplier No. C1306400100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Acetate Anhydrous ACS, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Specifications

Boiling Point 123°C
Melting Point 324°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Percent Purity ≥99%
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.528
Chemical Name or Material Sodium Acetate Anhydrous ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.