Learn More
Sodium Acetate + water in deuterium oxide, 99.9 atom D LC-NMR Reference Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Density | 1.104 g/mL (at 25°C) |
| Concentration | 50 mM in D 2 O (99.9 atom% D) |
| MDL Number | MFCD01094741 |
| Boiling Point | 102°C |
| Synonym | Sodium acetate solution |
| Type | NMR Reference Standard |
| Quantity | 1 Ea. |
| Grade | LC-NMR Reference Standard |
| Concentration or Composition (by Analyte or Components) | water, 1% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.