Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Aluminate, Technical, Spectrum™ Chemical
SDP

Catalog No. 18606291 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-291 500 g Poly Bottle
18-606-292 2.5 kg Poly Bottle
18-606-293 12 kg Poly Pail
18-606-294 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-291 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1120500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

S1120, 1302-42-7, Na2O·Al2O3·3H2O

Spectrum™ Chemical Sodium Aluminate, Technical is an inorganic chemical that is used as in water softening systems, as a coagulant aid to improve flocculation, and for removing dissolved silica and phosphates. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 1302-42-7
Molecular Formula AlNaO2
Molecular Weight (g/mol) 81.97
InChI Key IYJYQHRNMMNLRH-UHFFFAOYSA-N
IUPAC Name sodium dioxoalumanuide
SMILES [Na+].O=[Al-]=O

Specifications

CAS Max % 1
Quantity 500 g
UN Number UN2812
Grade Technical
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.