Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium anthraquinone-2-sulfonate

Catalog No. 743038500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-303-8500G 500 g
74-303-850G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74-303-8500G Supplier Sigma Aldrich Supplier No. 743038500G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium anthraquinone-2-sulfonate

Specifications

CAS 131-08-8
Molecular Formula C14H7NaO5S
Linear Formula C14H7NaO5S
MDL Number MFCD00001229
Quantity 500 g
Synonym 9,10-Dihydro-9,10-dioxo-2-anthracenesulfonic acid sodium salt; Anthraquinone-2-sulfonic acid sodium salt
Molecular Weight (g/mol) 310.26
Percent Purity ≥98% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.