Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Arsenate, Heptahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18613260 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
10 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-260 100 g Poly Bottle
18-613-259 25 g Poly Bottle
18-613-261 250 g Poly Bottle
18-613-262 1 kg Poly Bottle
18-613-263 10 kg Poly Pail
18-613-264 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-613-260 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1130100GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10048-95-0

Specifications

CAS Max % 1
Quantity 100 g
Percent Purity 98 to 102%
Grade ACS
Packaging Poly Bottle
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.